Tetraethyl orthosilicate (TEOS)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:78-10-4
:
Formula:Si(OC₂H₅)₄
MDL Number:MFCD00009062
:208.33 g/mol
Boiling Pt:168 °C
Density:0.934 g/cm³ (25 °C)
Flash Pt:48 °C
Storage Temperature:Ambient
EINECS:201-083-8
UN:1292
ADR:3,III
:GHS02
:GHS07
Frequently Bought Together