Auftragserfassung
Switzerland
ContactUsLinkComponent
6-Methylpyrazin-2-ol 98%
6-Methylpyrazin-2-ol 98%
Catalog # APOSOR40069-25G
Supplier:  Apollo Scientific
CAS Number:  
6-Methylpyrazin-2-ol 98%
Catalog # APOSOR40069-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Packung:
    Bottle
  • VE:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :110.1139 g/mol
Storage Temperature:Raumtemperatur
CAS Number:20721-18-0
Formula:C₅H₆N₂O
MDL Number:MFCD08705761