
1-Benzhydrylazetidin-3-ol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:18621-17-5
:
Formula:C₁₆H₁₇NO
MDL Number:MFCD00205109
:239.32 g/mol
Melting Pt:106…111 °C
Storage Temperature:Ambient
UN:3259
ADR:8,III
:GHS05
Frequently Bought Together