Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

D(-)-Isoleucine

Supplier:   Apollo Scientific
D(-)-Isoleucine
Supplier:  Apollo Scientific
Certificates

5 Options Available Below

Product Details & Documents
Synonyms:  (2R,3R)-(-)-Isoleucine, H-D-Ile-OH
  • CAS Number:319-78-8
  • Formula:H₃CCH₂CH(CH₃)CH(NH₂)COOH
  • MDL Number:MFCD00064221
  • Molecular Weight:131.17 g/mol
  • Melting Pt:265…274 °C
  •  :Density: 1.224 g/cm³ (20 °C)
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - SFr. 53.70
    SFr. 53.70
    1 * 100 mg
       SFr. 53.70

    Specifications:
    New Product
    New Product
  • Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - SFr. 91.80
    SFr. 91.80
    1 * 250 mg
       SFr. 91.80

  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - SFr. 246.00
    SFr. 246.00
    1 * 1 g
       SFr. 246.00

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - SFr. 856.00
    SFr. 856.00
    1 * 5 g
       SFr. 856.00

  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - SFr. 1’600.00
    SFr. 1’600.00
    1 * 25 g
       SFr. 1’600.00