Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Chloro-5-methylphenylboronic acid
3-Chloro-5-methylphenylboronic acid 98%
3-Chloro-5-methylphenylboronic acid 98%
APOSOR3621-1G
:
Apollo Scientific
:
3-Chloro-5-methylphenylboronic acid 98%
APOSOR3621-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
235…238 °C
:
170.40 g/mol
:
GHS07
:
Refrigerator
:
913836-14-3
:
C₇H₈BClO₂
:
MFCD09027241