2,3-Dihydro-1H-inden-1-amine hydrochloride 98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 1-Aminoindane hydrochloride
CAS Number:70146-15-5
:
Formula:C₉H₁₂ClN
MDL Number:MFCD00054294
Melting Pt:206…210 °C
:169.6513 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together