Order Entry
Switzerland
ContactUsLinkComponent
Ethyl-4-cyanobenzoate
Ethyl-4-cyanobenzoate
Catalog # APOSOR350177-25G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl-4-cyanobenzoate
Catalog # APOSOR350177-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :175.19 g/mol
Storage Temperature:Ambient
CAS Number:7153-22-2
Formula:C₁₀H₉NO₂
MDL Number:MFCD00001824