Order Entry
Switzerland
ContactUsLinkComponent
2,2,2-Trichloro-1-[4-(1-hydroxypropyl)-1H-pyrrol-2-yl]-1-ethanone 95%
2,2,2-Trichloro-1-[4-(1-hydroxypropyl)-1H-pyrrol-2-yl]-1-ethanone 95%
Catalog # APOSOR33491-1G
Supplier:  Apollo Scientific
CAS Number:  
2,2,2-Trichloro-1-[4-(1-hydroxypropyl)-1H-pyrrol-2-yl]-1-ethanone 95%
Catalog # APOSOR33491-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :270.54 g/mol
Storage Temperature:Ambient
CAS Number:1135283-05-4
Formula:C₉H₁₀Cl₃NO₂
MDL Number:MFCD11840981