

3-Chloro-N,N-diethylpropan-1-amine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:104-77-8
:
Formula:C₇H₁₆ClN
MDL Number:MFCD00039385
:149.66 g/mol
Boiling Pt:169…171 °C/760mmHg
Density:0.89 g/cm³ (25 °C)
Storage Temperature:Refrigerator
UN:1992
ADR:3+6.1,III
:GHS02
:GHS06
Frequently Bought Together