Methyl-4-formyl-2,5-dimethyl-1H-pyrrole-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:175205-91-1
:
Formula:C₉H₁₁NO₃
MDL Number:MFCD00203870
:181.19 g/mol
Melting Pt:179…181 °C
Storage Temperature:Ambient
Frequently Bought Together