
(2-Morpholinophenyl)methanol
Catalog # APOSOR23384-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
Melting Pt:54…57 °C
:193.240 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:465514-33-4
Formula:C₁₁H₁₅NO₂
MDL Number:MFCD03086181