2-Bromo-1-(pyridin-4-yl)ethanone hydrobromide
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 4-(Bromoacetyl)pyridine hydrobromide
CAS Number:5349-17-7
:
Formula:C₇H₇Br₂NO
MDL Number:MFCD02681893
Melting Pt:194…196 °C
:280.95 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together