Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

O411 98+%

New Product
Supplier:   Apollo Scientific
O411 98+%
Supplier:  Apollo Scientific
New Product
Certificates

4 Options Available Below

Product Details & Documents
Synonyms:  4-[(4-Methoxyphenyl)methoxy]benzeneacetonitrile, 2-(4-((4-Methoxybenzyl)oxy)phenyl)acetonitrile, O4I1
  • CAS Number:175135-47-4
  • Formula:C₁₆H₁₅NO₂
  • MDL Number:MFCD00067778
  • Molecular Weight:253.30 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - SFr. 217.00
    SFr. 217.00
    1 * 100 mg
       SFr. 217.00

    Specifications:
    New Product
    New Product
  • Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - SFr. 369.00
    SFr. 369.00
    1 * 250 mg
       SFr. 369.00

  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - SFr. 996.00
    SFr. 996.00
    1 * 1 g
       SFr. 996.00

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - SFr. 3’490.00
    SFr. 3’490.00
    1 * 5 g
       SFr. 3’490.00

    Specifications:
    New Product
    New Product