
3-Aminopiperidine-2,6-dione hydrochloride 97%
:24666-56-6
:
:C5H9ClN2O2
:MFCD11042437
:164.59 g/mol
:245 °C
:GHS07
:Ambient
Frequently Bought Together
Chlorobenzene 99+%, pure
: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
: Thermo Fisher Scientific
Chloroform
: Roth Carl
Isoamyl alcohol 98%, pure
: Thermo Fisher Scientific
Dichloromethane 99.6% stabilised ACS
: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
(±)-2-Butanol ≥99.5% (by GC), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Acetone 99.8% for HPLC
: Thermo Fisher Scientific