Order Entry
Switzerland
Orders LinkContactUsLinkComponent
D-(-)-Penicillamine

D-(-)-Penicillamine

Irritant
Supplier:   Apollo Scientific
D-(-)-Penicillamine
Supplier:  Apollo Scientific
Certificates
D-(-)-Penicillamine

6 Options Available Below

Product Details & Documents
Synonyms:  H-D-Pen-OH, (S)-2-Amino-3-mercapto-3-methylbutanoic acid, Penicillamine
  • CAS Number:52-67-5
  • Formula:C₅H₁₁NO₂S
  • MDL Number:MFCD00064302
  • Molecular Weight:149.2 g/mol
  • Melting Pt:∼ 212 °C (dec.)
  • Melting Pt:ca 212 °C (dec.)
  • Storage Temperature:Refrigerator
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - SFr. 26.00
    SFr. 26.00
    1 * 1 g
       SFr. 26.00

    Specifications:
    New Product
    New Product
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - SFr. 41.60
    SFr. 41.60
    1 * 5 g
       SFr. 41.60

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - SFr. 108.00
    SFr. 108.00
    1 * 25 g
       SFr. 108.00

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - SFr. 193.00
    SFr. 193.00
    1 * 100 g
       SFr. 193.00

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - SFr. 790.00
    SFr. 790.00
    1 * 500 g
       SFr. 790.00

  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - SFr. 3’790.00
    SFr. 3’790.00
    1 * 2,5 kg
       SFr. 3’790.00

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.