Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl (E)-4-chloro-3-methoxy-2-butenoate
Methyl (E)-4-chloro-3-methoxy-2-butenoate 95%
Methyl (E)-4-chloro-3-methoxy-2-butenoate 95%
APOSOR17691-25G
:
Apollo Scientific
:
Methyl (E)-4-chloro-3-methoxy-2-butenoate 95%
APOSOR17691-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
164.59 g/mol
:
30 °C
:
1.21 g/cm³ (25 °C)
:
Ambient
:
1759
:
8,III
:
GHS05
:
GHS07
:
110104-60-4
:
C₆H₉ClO₃
:
MFCD00071562