tert-Butyl-3-(hydroxymethyl)-2-methyl-1H-indole-1-carboxylate 98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: tert-Butyl 3-(hydroxymethyl)-2-methyl-1H-indole-1-carboxylate, 3-(Hydroxymethyl)-2-methyl-1H-indole, N-BOC protected
CAS Number:914349-13-6
:
Formula:C₁₅H₁₉NO₃
MDL Number:MFCD05864723
:261.3163 g/mol
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together