Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(4-Chlorophenyl)-N-methylmethanamine
1-(4-Chlorophenyl)-N-methylmethanamine
1-(4-Chlorophenyl)-N-methylmethanamine
1-(4-Chlorophenyl)-N-methylmethanamine
APOSOR16428-5G
:
Apollo Scientific
:
1-(4-Chlorophenyl)-N-methylmethanamine
APOSOR16428-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
155.6247 g/mol
:
77…78 °C/1mmHg
:
Ambient
:
201-175-3
:
3265
:
8,III
:
GHS05
:
GHS07
:
104-11-0
:
C₈H₁₀ClN
:
MFCD00018749