Order Entry
Switzerland
ContactUsLinkComponent
4-Bromophenyl phenyl ether (BDE No. 3)
4-Bromophenyl phenyl ether (BDE No. 3)
Catalog # APOSOR16402-25G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromophenyl phenyl ether (BDE No. 3)
Catalog # APOSOR16402-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 4-Bromodiphenyl ether, 1-Bromo-4-phenoxybenzene
 :
Boiling Pt:305 °C
Density:1.423 g/cm³ (25 °C)
Flash Pt:>113 °C
ADR:9+EHS,III
 :249.11 g/mol
UN:3082
 :GHS05
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:101-55-3
Formula:C₁₂H₉BrO
MDL Number:MFCD00000094