2-(2,5-Dioxopyrrolidin-1-yl)benzoic acid
Catalog # APOSOR16148-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:219.2 g/mol
Storage Temperature:Ambient
CAS Number:80022-74-8
Formula:C₁₁H₉NO₄
MDL Number:MFCD00022590