
6-Phenyl-3(2H)-pyridazinone
Catalog # APOSOR14974-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:172.19 g/mol
Melting Pt:201…204 °C
EINECS:218-505-1
:GHS07
Storage Temperature:Ambient
CAS Number:2166-31-6
Formula:C₁₀H₈N₂O
MDL Number:MFCD00114953