

5-Hexen-1-ol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:821-41-0
:
Formula:C₆H₁₂O
MDL Number:MFCD00002981
:100.16 g/mol
Boiling Pt:152…155 °C
Melting Pt:< –20 °C
Density:0.834 g/cm³ (25 °C)
Flash Pt:47 °C
Storage Temperature:Ambient
EINECS:212-477-4
UN:2924
ADR:3+8,III
:GHS02
:GHS05
Frequently Bought Together