Order Entry
Switzerland
ContactUsLinkComponent
5-Methoxy-1H-indole-3-carboxylic acid 98%
5-Methoxy-1H-indole-3-carboxylic acid 98%
Catalog # APOSOR13726-5G
Supplier:  Apollo Scientific
CAS Number:  
5-Methoxy-1H-indole-3-carboxylic acid 98%
Catalog # APOSOR13726-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :191.19 g/mol
Storage Temperature:Ambient
CAS Number:10242-01-0
Formula:C10H9NO3
MDL Number:MFCD03265451