

2-Methylpyridin-3-ol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1121-25-1
:
Formula:C₆H₇NO
MDL Number:MFCD00082538
:109.13 g/mol
Melting Pt:168…169 °C
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together