
4-Propoxy-3-methylphenylboronsäure
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:279262-88-3
:
Formula:C₁₀H₁₅BO₃
MDL Number:MFCD06659872
:194.0400 g/mol
Melting Pt:154…156 °C
:GHS07
Storage Temperature:Kühlschrank
Frequently Bought Together