4-Chloro-m-anisidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:13726-14-2
:
Formula:C₇H₈ClNO
MDL Number:MFCD00672967
:157.6 g/mol
Melting Pt:78…81 °C
UN:2233
ADR:6.1,III
:GHS06
:GHS08
Storage Temperature:Ambient
Frequently Bought Together