Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Auftragserfassung
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Benzyloxy-2-methylphenylboronsäure
4-Benzyloxy-2-methylphenylboronsäure
4-Benzyloxy-2-methylphenylboronsäure
APOSOR10356-25G
:
Apollo Scientific
:
4-Benzyloxy-2-methylphenylboronsäure
APOSOR10356-25G
:
Apollo Scientific
:
Packung:
Bottle
VE:
25 g
Seq:
3308
:
:
242.08 g/mol
:
154…158 °C
:
GHS07
:
Kühlschrank
:
847560-49-0
:
C₁₄H₁₅BO₃
:
MFCD03788404