Order Entry
Switzerland
Orders LinkContactUsLinkComponent
7-Hydroxyindole 97%

7-Hydroxyindole 97%

Irritant
Supplier:   Apollo Scientific
7-Hydroxyindole 97%
Supplier:  Apollo Scientific
Certificates

2 Options Available Below

Product Details & Documents
  • CAS Number:2380-84-9
  • Formula:C₈H₇NO
  • MDL Number:MFCD00152102
  • Molecular Weight:133.15 g/mol
  • Melting Pt:84…88 °C
  • Storage Temperature:Freezer
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 139.00 SFr.
    139.00 SFr.
    1 * 1 g
       139.00 SFr.

    Specifications:
    More
    Specifications
    Cat. No.
    APOSOR10209-1G
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 416.00 SFr.
    416.00 SFr.
    1 * 5 g
       416.00 SFr.

    Specifications:
    More
    Specifications
    Cat. No.
    APOSOR10209-5G
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.