Order Entry
Switzerland
ContactUsLinkComponent
1,8-Naphthyridine-2-carboxylic acid 97%
Irritant
1,8-Naphthyridine-2-carboxylic acid 97%
Catalog # APOSOR1008-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,8-Naphthyridine-2-carboxylic acid 97%
Catalog # APOSOR1008-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :174.16 g/mol
Melting Pt:220 °C (dec.)
 :GHS07
Storage Temperature:Ambient
CAS Number:215523-34-5
Formula:C₉H₆N₂O₂
MDL Number:MFCD00202900