Order Entry
Switzerland
Orders LinkContactUsLinkComponent
L(+)-Cysteine

L(+)-Cysteine

Irritant
New Product
Supplier:   Apollo Scientific
L(+)-Cysteine
Supplier:  Apollo Scientific
New Product
Certificates

6 Options Available Below

Product Details & Documents
Synonyms:  H-Cys-OH, L-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid
  • CAS Number:52-90-4
  • Formula:HSCH₂CH(NH₂)CO₂H
  • MDL Number:MFCD00064306
  • Molecular Weight:121.16 g/mol
  • Melting Pt:240 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-158-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - SFr. 26.00
    SFr. 26.00
    1 * 100 g
       SFr. 26.00

  • Bottle
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - SFr. 34.70
    SFr. 34.70
    1 * 250 g
       SFr. 34.70

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - SFr. 60.70
    SFr. 60.70
    1 * 500 g
       SFr. 60.70

    Specifications:
    New Product
    New Product
  • Bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - SFr. 113.00
    SFr. 113.00
    1 * 1 kg
       SFr. 113.00

  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - SFr. 555.00
    SFr. 555.00
    1 * 5 kg
       SFr. 555.00

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 kg
    1 * 10 kg - SFr. 1’100.00
    SFr. 1’100.00
    1 * 10 kg
       SFr. 1’100.00

    Specifications:
    New Product
    New Product