
4-(4-Chlorophenyl)-1-butene
Catalog # APOSOR01895-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:166.65 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:3047-24-3
Formula:C₁₀H₁₁Cl
MDL Number:MFCD01319548