
Ac-α-benzyl-4,6-O-benzylidene muramic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2862-03-5
:
Formula:C₂₅H₂₉NO₈
MDL Number:MFCD00057299
:471.506 g/mol
Melting Pt:220…222 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together