Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Heim
Oxalic acid dihydrate
Oxalic acid dihydrate ACS
Oxalic acid dihydrate ACS
APOSIN2794-1KG
:
Apollo Scientific
:
Oxalic acid dihydrate ACS
APOSIN2794-1KG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 kg
Seq:
4024
:
:
126.07 g/mol
:
101…102 °C
:
1.65 g/cm³ (25 °C)
:
Ambient
:
607-634-3
:
3261
:
8,III
:
GHS05
:
GHS07
:
6153-56-6
:
HO₂CCO₂H·2H₂O
:
MFCD00149102