SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Manganese (II) chloride tetrahydrate
Manganese(II) chloride tetrahydrate 99%
Manganese(II) chloride tetrahydrate 99%
APOSIN2521-100G
:
Apollo Scientific
:
Manganese(II) chloride tetrahydrate 99%
APOSIN2521-100G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 g
Seq:
3401
:
:
197.9 g/mol
:
106 °C
:
58 °C
:
2.01 g/cm³ (25 °C)
:
GHS07
:
Ambient
:
13446-34-9
:
MnCl₂·4H₂O
:
MFCD00149792