Rifampicin 99% approx
:13292-46-1
:
:C₄₃H₅₈N₄O₁₂
:MFCD00151389
:822.95 g/mol
:183…194 °C
:236-312-0
:GHS07
:Freezer
Frequently Bought Together
Transfer pipettes
: RATIOLAB
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Agar, powder for bacteriology
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF