Order Entry
Switzerland
Orders LinkContactUsLinkComponent
m-PEG4-amine

m-PEG4-amine

New Product
Supplier:   Apollo Scientific
m-PEG4-amine
Supplier:  Apollo Scientific
New Product
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  3,6,9,12-Tetraoxatridecanamine
  • CAS Number:85030-56-4
  • Formula:C₉H₂₁NO₄
  • Molecular Weight:207.27 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - SFr. 83.20
    SFr. 83.20
    1 * 1 g
       SFr. 83.20

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - SFr. 411.00
    SFr. 411.00
    1 * 5 g
       SFr. 411.00

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - SFr. 1’890.00
    SFr. 1’890.00
    1 * 25 g
       SFr. 1’890.00

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.