


Phthalaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:643-79-8
:
Formula:C₈H₆O₂
MDL Number:MFCD00003335
:134.13 g/mol
Boiling Pt:83 °C/0.8mmHg
Melting Pt:53…57 °C
Density:1.13 g/cm³ (25 °C)
Flash Pt:132 °C
Storage Temperature:Refrigerator
EINECS:211-402-2
UN:2923
ADR:8+6.1,II
:GHS05
:GHS06
:GHS09
Frequently Bought Together