Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Agar

Agar

Irritant
Supplier:   Apollo Scientific
Agar
Supplier:  Apollo Scientific
Certificates

3 Options Available Below

Product Details & Documents

A purified agar with a high gel strength and clarity for tissue culture and microbiological work. Minimal conc. of 5.0g/L to obtain solid gel.

Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Storage Temperature:Ambient
  • EINECS:232-658-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - SFr. 86.60
    SFr. 86.60
    1 * 500 g
       SFr. 86.60

  • Bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - SFr. 148.00
    SFr. 148.00
    1 * 1 kg
       SFr. 148.00

  • Bottle
    5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - SFr. 650.00
    SFr. 650.00
    1 * 5 kg
       SFr. 650.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.