Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Quercetin dihydrate

Quercetin dihydrate

Supplier:   Apollo Scientific
Quercetin dihydrate
Supplier:  Apollo Scientific
Certificates
Quercetin dihydrate

2 Options Available Below

Product Details & Documents
Synonyms:  2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one dihydrate
  • CAS Number:6151-25-3
  • Formula:C₁₅H₁₄O₉
  • MDL Number:MFCD00149487
  • Molecular Weight:338.268 g/mol
  • Melting Pt:315 °C (dec.)
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - SFr. 104.00
    SFr. 104.00
    1 * 25 g
       SFr. 104.00

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - SFr. 381.00
    SFr. 381.00
    1 * 100 g
       SFr. 381.00