


Selumetinib free base 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:606143-52-6
:
Formula:C₁₇H₁₅BrClFN₄O₃
MDL Number:MFCD11977472
:457.7 g/mol
Melting Pt:208…214 °C
:GHS05
:GHS07
:GHS08
Storage Temperature:Freezer
Frequently Bought Together