
Sodium-3-[ethyl(m-tolyl)amino]-2-hydroxy-1-propanesulphonate hydrate ≥98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:82692-93-1
:
Formula:C₁₂H₁₉NO₄S
MDL Number:MFCD00040641
:295.33 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together