Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Digitonin

Digitonin

Toxic
Supplier:   Apollo Scientific
Digitonin
Supplier:  Apollo Scientific
Certificates

3 Options Available Below

Product Details & Documents

Non-ionic detergent used to permeabilize cell membranes.

Synonyms:  Digitonin
  • CAS Number:11024-24-1
  • Formula:C₅₆H₉₂O₂₉
  • MDL Number:MFCD00077729
  • Molecular Weight:1229.312 g/mol
  • Storage Temperature:Ambient
  • EINECS:234-255-6
  • UN:3462
  • ADR:6.1,III

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 65.90 SFr.
    65.90 SFr.
    1 * 250 mg
       65.90 SFr.

  • Bottle
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 113.00 SFr.
    113.00 SFr.
    1 * 500 mg
       113.00 SFr.

  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 191.00 SFr.
    191.00 SFr.
    1 * 1 g
       191.00 SFr.