
D-(+)-Glucose-[1-¹³C] 99% min (Isotope 13C 99% )
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:40762-22-9
:
Formula:C₆H₁₂O₆
MDL Number:MFCD00037545
Melting Pt:146…152 °C
:GHS07
Storage Temperature:Freezer
Frequently Bought Together