Order Entry
Switzerland
ContactUsLinkComponent
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)
Catalog # APOSBICL207-100MG
Supplier:  Apollo Scientific
CAS Number:  
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)
Catalog # APOSBICL207-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :436.37 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:92921-24-9
Formula:C₁₆H₁₈N₂O₉S
MDL Number:MFCD00054978