
EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)]
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:70539-42-3
:
Formula:C₁₈H₂₀N₂O₁₂
MDL Number:MFCD00049057
:456.36 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together