
DSS (Disuccinimidyl suberate)
Catalog # APOSBICL100-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:368.34 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:68528-80-3
Formula:C₁₆H₂₀N₂O₈
MDL Number:MFCD00049059