Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Methyl-α-D-glucopyranoside

Methyl-α-D-glucopyranoside

Bottle, 500 g
Catalog # APOSBIB6105-500G
Irritant
Supplier:   Apollo Scientific
Methyl-α-D-glucopyranoside
Catalog # APOSBIB6105-500G
Supplier:  Apollo Scientific
CAS Number:  97-30-3
Methyl-α-D-glucopyranoside

Bottle, 500 g

CAS Number:  97-30-3
Price
SFr. 45.10
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  α-D-Methylglucoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol
  • CAS Number:97-30-3
  • Formula:C₇H₁₄O₆
  • MDL Number:MFCD00064086
  • Molecular Weight:194.18 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.