Caffeic acid
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid
- CAS Number:331-39-5
- Formula:C₉H₈O₄
- MDL Number:MFCD00004392
- Molecular Weight:180.16 g/mol
- Melting Pt:211…213 °C (dec.)
- Storage Temperature:Ambient
- EINECS:206-361-2
4 Options Available Below
Product Details & Documents
Synonyms: 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid
- CAS Number:331-39-5
- Formula:C₉H₈O₄
- MDL Number:MFCD00004392
- Molecular Weight:180.16 g/mol
- Melting Pt:211…213 °C (dec.)
- Storage Temperature:Ambient
- EINECS:206-361-2
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



