L(+)-Arginine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:74-79-3
:
Formula:C₆H₁₄N₄O₂
MDL Number:MFCD00002635
:174.2 g/mol
Melting Pt:222…226 °C (dec.)
EINECS:200-811-1
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together