Actinomycin D ≥99%
Supplier: Apollo Scientific
Jump to:
About this item
Antineoplastic antibiotic
- Purity: ≥99%
- CAS Number:50-76-0
- Formula:C₆₂H₈₆N₁₂O₁₆
- MDL Number:MFCD00005033
- Molecular Weight:1255.5 g/mol
- Melting Pt:251…253 °C
- Storage Temperature:Freezer
- EINECS:200-063-6
- UN:3462
- ADR:6.1,II
6 Options Available Below
Product Details & Documents
Antineoplastic antibiotic
- Purity: ≥99%
- CAS Number:50-76-0
- Formula:C₆₂H₈₆N₁₂O₁₆
- MDL Number:MFCD00005033
- Molecular Weight:1255.5 g/mol
- Melting Pt:251…253 °C
- Storage Temperature:Freezer
- EINECS:200-063-6
- UN:3462
- ADR:6.1,II
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



